CHAPS, Electrophoresis Grade, Thermo Scientific Chemicals
CHAPS, Electrophoresis Grade, Thermo Scientific Chemicals
CHAPS, Electrophoresis Grade, Thermo Scientific Chemicals
CHAPS, Electrophoresis Grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals
CHAPS, Electrophoresis Grade, Thermo Scientific Chemicals
CHAPS, 3-[(3-cholamidopropyl)dimethylammonio]-1-propanesulfonate, CAS # 75621-03-3, is a non-denaturing zwitterionic detergent that can be used for solubilizing membrane proteins.
Have Questions?
Catalog NumberQuantity
ALFJ67359.065 g
Catalog number ALFJ67359.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS822-16-2
IUPAC Namesodium octadecanoate
Molecular FormulaC18H35NaO2
InChI KeyRYYKJJJTJZKILX-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCCCCCCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
CommentFree Fatty Acid (FFA): ≤3.0%
CommentMelting Point: ≥200°C
Loss on Drying≤3.0%
CHAPS is often used as a detergent in the solubilization and purification of membrane proteins for several advantageous reasons. CHAPS detergent is non-denaturing to membrane proteins, can solubilize proteins, disaggregate protein-protein interactions. CHAPS is also useful in ion exchange chromatography and isoelectric focusing as it is zwitterionic and does not exhibit a net charge between pH 2 to 12. It is suitable for isolation of membrane proteins with the following features such as sufficient protein solubilization capability, no denaturing or inactivation of proteins, no interference with protein activities, no precipitation at 4°C, appropriate critical micelle concentrations (CMC) and micelle size, no absorption in the UV region, no toxicity and availability of detergent detection methods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• CHAPS (3-((3-cholamidopropyl) dimethylammonio)-1-propanesulfonate) is a zwitterionic non-denaturing detergent
• It is suitable for applications which require protection of native protein conformation
Applications
• CHAPS can be used for the solubilization and purification of membrane proteins
• It is also used in conjunction with nonionic detergents like Triton X-100
• It can be used as a non-denaturing zwitterionic detergent for the study of membrane chemistry
RUO – Research Use Only
General References:
  1. Rodi, P. M.; Gianello, M. D. B.; Corregido, M. C.; Gennaro, A. M. Comparative study of the interaction of CHAPS and Triton X-100 with the erythrocyte membrane. Biochim. Biophys. Acta 2014, 1838 (3), 859-866.
  2. Ghosh, A.; Saha, R.; Saha, B. Effect of CHAPS and CPC micelles on Ir(III) catalyzed Ce(IV) oxidation of aliphatic alcohols at room temperature and pressure. J. Mol. Liq. 2014, 196, 223-237.