Anhydrotetracycline hydrochloride, Thermo Scientific Chemicals
Anhydrotetracycline hydrochloride, Thermo Scientific Chemicals
Anhydrotetracycline hydrochloride, Thermo Scientific Chemicals
Anhydrotetracycline hydrochloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Anhydrotetracycline hydrochloride, Thermo Scientific Chemicals
Anhydrotetracycline hydrochloride, CAS # 13803-65-1, is a tetracycline derivative showing antibacterial activity commonly used to control tetracycline-dependent gene expression systems in bacteria.
Have Questions?
Catalog NumberQuantity
ALFJ66688.MA10 mg
Catalog number ALFJ66688.MA
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 mg
Request bulk or custom format
Chemical Identifiers
CAS108-91-8
IUPAC Namecyclohexanamine
Molecular FormulaC6H13N
InChI KeyPAFZNILMFXTMIY-UHFFFAOYSA-N
SMILESNC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=98.5 %
Refractive index1.4580 to 1.4600 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
View more
Anhydrotetracycline hydrochloride acts as a useful effector of tetracycline-dependent gene expression in bacterial expression systems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Anhydrotetracycline hydrochloride is a degradation product derived from tetracycline hydrochloride. It acts as an effector of tetracycline-dependent gene expression in bacterial expression systems. In this system the transcription of genes is reversibly turned on/off in the presence of several compounds including, anhydrotetracycline hydrochloride.
• This compound poorly binds to the 30S ribosomal subunit compared with its precursor, tetracycline.
Applications
• Anhydrotetracycline hydrochloride is commonly used in all fields of biology for control of transcription or translation processes
• This compound has been used in laboratory experiments and shows lower antibacterial activity against Escherichia coli compared with tetracycline.
RUO – Research Use Only
General References:
  1. Baumschlager, A.; Rullan, M.; Khammash, M. Exploiting natural chemical photosensitivity of anhydrotetracycline and tetracycline for dynamic and setpoint chemo-optogenetic control. Nature Communications. 2020, 11, 3834.
  2. Syed Laik Ali.; Thomas Strittmatter. Separation and quantitative determination of tetracycline, epitetracycline, epianhydrotetracycline and anhydrotetracycline by high-performance liquid chromatography. International Journal of Pharmaceutics. 1978, 1, (3), 185-188.
  3. Lloyd, P.B.; Carol C. Cornford. A thin-layer chromatographic limit test for the detection of anhydrotetracycline and 4-epi-anhydrotetracycline in tetracycline. Journal of Chromatography A. 1970, 53, (2), 403-405.
  4. Xie, L.; Wang, C.; Chen, M.; Jin, B.; Zhou, R.; Huang, Y.; Hameed, S.; Ying, Y. Temperature-dependent terahertz vibrational spectra of tetracycline and its degradation products. Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy. 2019, 5, 222, 117179.