4-Methylumbelliferyl beta-D-glucopyranoside, 99%, Thermo Scientific Chemicals
4-Methylumbelliferyl beta-D-glucopyranoside, 99%, Thermo Scientific Chemicals
4-Methylumbelliferyl beta-D-glucopyranoside, 99%, Thermo Scientific Chemicals
4-Methylumbelliferyl beta-D-glucopyranoside, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Methylumbelliferyl beta-D-glucopyranoside, 99%, Thermo Scientific Chemicals
CAS: 18997-57-4 | C16H18O8 | 338.312 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ66630.065 g
Catalog number ALFJ66630.06
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Methylumbelliferyl beta-D-glucopyranoside
CAS18997-57-4
Melting Point210°C to 212°C
ColorWhite
Recommended StorageStore at -20°C