Triton∣r X-100, Electrophoresis Reagent
Triton∣r X-100, Electrophoresis Reagent
Triton∣r X-100, Electrophoresis Reagent
Triton∣r X-100, Electrophoresis Reagent
Thermo Scientific Chemicals
Triton∣r X-100, Electrophoresis Reagent
CAS: 9002-93-1 | C16H26O2, C16H26O2 | 250.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2500 mL
Catalog number ALFJ66624.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS553-03-7
IUPAC Name1,2,3,4-tetrahydroquinolin-2-one
Molecular FormulaC9H9NO
InChI KeyTZOYXRMEFDYWDQ-UHFFFAOYSA-N
SMILESO=C1CCC2=CC=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)163-171°C
Appearance (Color)White to cream to pale brown
FormPowder
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
Ambient temperatures