Kainic acid monohydrate, 96%
Kainic acid monohydrate, 96%
Kainic acid monohydrate, 96%
Kainic acid monohydrate, 96%
Thermo Scientific Chemicals
Kainic acid monohydrate, 96%
CAS: 58002-62-3 | C10H17NO5, C10H15NO4 | 231.25 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ66590.MA10 mg
Catalog number ALFJ66590.MA
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 mg
Request bulk or custom format
Chemical Identifiers
CAS2409-55-4
IUPAC Name2-tert-butyl-4-methylphenol
Molecular FormulaC11H16O
InChI KeyIKEHOXWJQXIQAG-UHFFFAOYSA-N
SMILESCC1=CC=C(O)C(=C1)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to light beige
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Crystalline mass
Melting point50°C to 53°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
2°C to 8°C