Laminarin, Thermo Scientific Chemicals
Laminarin, Thermo Scientific Chemicals
Laminarin, Thermo Scientific Chemicals
Laminarin, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Laminarin, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number ALFJ66193.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS115-41-3
IUPAC Name3,3-bis(3,4-dihydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular FormulaC19H14O7S
InChI KeyRRRCKIRSVQAAAS-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Solubility(0.1 % in water) Clear
Appearance (Color)Green to dark green to brown
Infrared spectrumConforms
UV spectrumComplies
UV Lambda max441 to 449 nm (in methanol)
View more
Laminarin is a storage glucan and is now used to study it’s interaction with laminarinase as its substrate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Laminarin is a storage glucan and is now used to study it’s interaction with laminarinase as its substrate.
Solubility
Soluble in dimethyl sulfoxide (slightly, heated). Slightly soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Hui Dai; Xiao-Qiang Han; Fang-Yuan Gong; Hongliang Dong; Peng-Fei Tu; and Xiao-Ming Gao. Structure elucidation and immunological function analysis of a novel β-glucan from the fruit bodies of Polyporus umbellatus (Pers.) Fries. Glycobiology, 2012,22(12), 1673-1683.
  2. Park, H.K., et al. Induction of apoptosis by laminarin, regulating the insulin-like growth factor-IR signaling pathways in HT-29 human colon cells. Int. J. Molec. Med, 2012,30(4), 734-738.