Ethidium Bromide, Electrophoresis Reagent, 98% (dry weight)
Ethidium Bromide, Electrophoresis Reagent, 98% (dry weight)
Ethidium Bromide, Electrophoresis Reagent, 98% (dry weight)
Ethidium Bromide, Electrophoresis Reagent, 98% (dry weight)
Thermo Scientific Chemicals
Ethidium Bromide, Electrophoresis Reagent, 98% (dry weight)
CAS: 1239-45-8 | C21H20BrN3, C21H20BrN3 | 394.32 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ66192.031 g
Catalog number ALFJ66192.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Assay98%
CAS1239-45-8
ColorRed to purple
Recommended StorageAmbient temperatures
EINECS Number214-984-6