Poloxamer 188, Thermo Scientific Chemicals
Poloxamer 188, Thermo Scientific Chemicals
Poloxamer 188, Thermo Scientific Chemicals
Poloxamer 188, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Poloxamer 188, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Catalog number ALFJ66087.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPoloxamer 188
CAS9003-11-6
ColorWhite
Quantity500 g
Molecular FormulaC5H10O2
View more
Poloxamer is a non-ionic detergent that protects cells from hydrodynamic damage. It can be also used for in vitro isolation of lymphocytes from diluted whole blood.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Poloxamer is a non-ionic detergent that protects cells from hydrodynamic damage. It can be also used for in vitro isolation of lymphocytes from diluted whole blood.
Solubility
Miscible with water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. Goppert TM.; Muller RH. Plasma protein adsorption of Tween 80- and poloxamer 188-stabilized solid lipid nanoparticles.. Journal of Drug Targeting. 2003, 11, (4), 225-231.
  2. Carr ME Jr.; Powers PL.; Jones MR. Effects of poloxamer 188 on the assembly, structure and dissolution of fibrin clots. Thrombosis and Haemostasis. 1991, 66, (5), 565-568.
Contents & Storage
Ambient temperatures