D-Glucose-6-phosphate disodium salt hydrate, 98%
D-Glucose-6-phosphate disodium salt hydrate, 98%
D-Glucose-6-phosphate disodium salt hydrate, 98%
D-Glucose-6-phosphate disodium salt hydrate, 98%
Thermo Scientific Chemicals
D-Glucose-6-phosphate disodium salt hydrate, 98%
CAS: 3671-99-6 | C6H11Na2O9P | 304.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALFJ66023.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialD-Glucose-6-phosphate disodium salt
CAS3671-99-6
ColorWhite
Recommended StorageStore at -20°C
EINECS Number222-938-1