5-Bromo-4-chloro-3-indolyl beta-D-glucoside, 99%, Thermo Scientific Chemicals
5-Bromo-4-chloro-3-indolyl beta-D-glucoside, 99%, Thermo Scientific Chemicals
5-Bromo-4-chloro-3-indolyl beta-D-glucoside, 99%, Thermo Scientific Chemicals
5-Bromo-4-chloro-3-indolyl beta-D-glucoside, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Bromo-4-chloro-3-indolyl beta-D-glucoside, 99%, Thermo Scientific Chemicals
Substrate for beta→-D-glucosidase
Have Questions?
Catalog NumberQuantity
ALFJ65677.ME500 mg
Catalog number ALFJ65677.ME
Quantity:
500 mg
Request bulk or custom format
Chemical Identifiers
CAS920-66-1
IUPAC Name1,1,1,3,3,3-hexafluoropropan-2-ol
Molecular FormulaC3H2F6O
InChI KeyBYEAHWXPCBROCE-UHFFFAOYSA-N
SMILESOC(C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.3 %
GC>=99.50 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Substrate for beta--D-glucosidase