8-Anilinonaphthalene-1-sulfonic acid, 95%, Thermo Scientific Chemicals
8-Anilinonaphthalene-1-sulfonic acid, 95%, Thermo Scientific Chemicals
8-Anilinonaphthalene-1-sulfonic acid, 95%, Thermo Scientific Chemicals
8-Anilinonaphthalene-1-sulfonic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
8-Anilinonaphthalene-1-sulfonic acid, 95%, Thermo Scientific Chemicals
Fluorescent probe for proteins on polyacrylamide gels
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number ALFJ65332.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS7782-89-0
IUPAC Namelithium(1+) azanide
Molecular FormulaH2LiN
InChI KeyAFRJJFRNGGLMDW-UHFFFAOYSA-N
SMILES[Li+].[NH2-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey
FormPowder (may contain black specks)
Assay (unspecified)≥94.0%
Fluorescent probe for proteins on polyacrylamide gels