8-Aminonaphthalene-2-sulfonic acid, 95%, Thermo Scientific Chemicals
8-Aminonaphthalene-2-sulfonic acid, 95%, Thermo Scientific Chemicals
8-Aminonaphthalene-2-sulfonic acid, 95%, Thermo Scientific Chemicals
8-Aminonaphthalene-2-sulfonic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
8-Aminonaphthalene-2-sulfonic acid, 95%, Thermo Scientific Chemicals
CAS: 119-28-8 | C10H9NO3S | 223.246 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALFJ65304.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1313-96-8
IUPAC Name[(dioxoniobio)oxy]dioxoniobium
Molecular FormulaNb2O5
InChI KeyZKATWMILCYLAPD-UHFFFAOYSA-N
SMILESO=[Nb](=O)O[Nb](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.