Cellulase, chromatographically purified, T. reesei
Cellulase, chromatographically purified, T. reesei
Thermo Scientific Chemicals
Cellulase, chromatographically purified, T. reesei
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number ALFJ64019.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS620-08-6
IUPAC Name4-methoxypyridine
Molecular FormulaC6H7NO
InChI KeyXQABVLBGNWBWIV-UHFFFAOYSA-N
SMILESCOC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥98.0%
CommentMay darken on storage
Refractive Index1.5165-1.5235 @ 20?C
Cellulase has been used to degrade cello-oligosaccharides into glucose and to study the biodegradability of bioabsorbable bacterial cellulose (BBC) material. It has been used in a study to assess the differential regulation of the cellulase transcription factors XYR1, ACE2, and ACE. It is also applied in the biodegradation of paper products such as foolscap paper, filter paper, newspaper and office paper as well as microcrystalline cellulose. It is added to detergents to improve cleansing properties.