Colcemid, 98+%
Colcemid, 98+%
Colcemid, 98+%
Colcemid, 98+%
Thermo Scientific Chemicals
Colcemid, 98+%
CAS: 477-30-5 | C21H25NO5, C21H25NO5 | 371.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
10 mg
Catalog number ALFJ63900.MF
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mg
Request bulk or custom format
Specifications
Quantity50 mg
DescriptionPale yellow
For Use With (Application)Microtubule modulator that depolymerizes microtubules and inactivates spindle fiber formation
Assay98+%
CAS477-30-5
View more
Microtubule modulator that depolymerizes microtubules and inactivates spindle fiber formation
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
2°C to 8°C