Colcemid, 98+%
Colcemid, 98+%
Colcemid, 98+%
Colcemid, 98+%
Thermo Scientific Chemicals
Colcemid, 98+%
CAS: 477-30-5 | C21H25NO5, C21H25NO5 | 371.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
50 mg
Catalog number ALFJ63900.MA
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 mg
Request bulk or custom format
Chemical Identifiers
CAS22767-49-3
IUPAC Namesodium pentane-1-sulfonate
Molecular FormulaC5H11NaO3S
InChI KeyROBLTDOHDSGGDT-UHFFFAOYSA-M
SMILES[Na+].CCCCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
Appearance (Color)White
Loss on drying=<2 % (1 g, 120°C)
Titration after Ion exchange>=98.0 % (on dry substance)
View more
Microtubule modulator that depolymerizes microtubules and inactivates spindle fiber formation
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
2°C to 8°C