Sodium phosphate, 0.2M buffer soln., pH 7.2
Sodium phosphate, 0.2M buffer soln., pH 7.2
Thermo Scientific Chemicals
Sodium phosphate, 0.2M buffer soln., pH 7.2
Have Questions?
Catalog NumberQuantity
ALFJ63816.K21 L
Catalog number ALFJ63816.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS551-08-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, yellow
Assay (GC)≥94.0% (sum of isomers)
Identification (FTIR)Conforms
Refractive Index1.5740-1.5800 @ 20?C
FormLiquid
Sodium phosphate is widely used in molecular biology, biochemistry, and chromatography. Useful in conjunction with Sodium Phosphate, Monobasic in the preparation of biological buffers. Used in many applications including the purification of antibodies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium phosphate is widely used in molecular biology, biochemistry, and chromatography. Useful in conjunction with Sodium Phosphate, Monobasic in the preparation of biological buffers. Used in many applications including the purification of antibodies.
Solubility
Fully miscible.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. Masters, C.L.; Simms, G.; Weinman, N.A.; Multhaup, G.; McDonald, B.L.; Beyreuther, K. Amyloid plaque core protein in Alzheimer disease and Down syndrome. Current Issue. 1985, 82, (12), 4245-4249.
  2. Volker M. Vogt. Purification and Further Properties of Single-Strand-Specific Nuclease from Aspergillus oryzae. European Journal of Biochemistry. 1973, 33, (1), 192-200.