Borate, 0.5M buffer soln., pH 8.0
Borate, 0.5M buffer soln., pH 8.0
Borate, 0.5M buffer soln., pH 8.0
Borate, 0.5M buffer soln., pH 8.0
Thermo Scientific Chemicals
Borate, 0.5M buffer soln., pH 8.0
CAS: 1330-43-4 | B4Na2O7 | 201.21 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ63742.AP500 mL
Catalog number ALFJ63742.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialBorate buffer soln.
CAS1330-43-4
Health Hazard 1H319-H360FD
Health Hazard 2GHS H Statement
H360
May damage fertility or the unborn child.
Health Hazard 3P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c