Domperidone, Thermo Scientific Chemicals
Domperidone, Thermo Scientific Chemicals
Domperidone, Thermo Scientific Chemicals
Domperidone, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Domperidone, Thermo Scientific Chemicals
Peripheral dopamine receptor antagonist that does not cross the blood-brain barrier
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
100 mg
500 mg
Catalog number ALFJ63681.MF
Quantity:
50 mg
Request bulk or custom format
Chemical Identifiers
CAS767-00-0
IUPAC Name4-hydroxybenzonitrile
Molecular FormulaC7H5NO
InChI KeyCVNOWLNNPYYEOH-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder
Melting Point (clear melt)110-115?C
Assay (GC)≥98.0%
Domperidone is used as a Peripheral dopamine receptor antagonist that does not cross the blood-brain barrier. The antidopaminergic effects of Domperidone have been correlated to antiemetic and gastroprokinetic action.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Domperidone is used as a Peripheral dopamine receptor antagonist that does not cross the blood-brain barrier. The antidopaminergic effects of Domperidone have been correlated to antiemetic and gastroprokinetic action.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only
General References:
  1. Pierre M. Laduron.; Josee E. Leysen. Domperidone, a specific in vitro dopamine antagonist, devoid of in vivo central dopaminergic activity. Biochemical Pharmacology. 1979, 28, (14), 2161-2165.
  2. Jai D. Kohli.; Dana Glock.; Leon I. Goldberg. Selective DA2 versus DA1 antagonist activity of domperidone in the periphery. European Journal of Pharmacology. 1983, 89, (1-2), 137-141.