Genistin, 99+%, Thermo Scientific Chemicals
Genistin, 99+%, Thermo Scientific Chemicals
Genistin, 99+%, Thermo Scientific Chemicals
Genistin, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Genistin, 99+%, Thermo Scientific Chemicals
Glucoside of genistein that inhibits protein tyrosine kinase
Have Questions?
Catalog NumberQuantity
ALFJ63445.MC100 mg
Catalog number ALFJ63445.MC
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialGenistin
CAS529-59-9
Melting Point258°C to 259°C
ColorBrown to Yellow
Recommended StorageStore at -20°C
View more
Glucoside of genistein that inhibits protein tyrosine kinase