Cytidine-5'-monophosphate disodium salt, 99+%
Cytidine-5'-monophosphate disodium salt, 99+%
Cytidine-5'-monophosphate disodium salt, 99+%
Cytidine-5'-monophosphate disodium salt, 99+%
Thermo Scientific Chemicals
Cytidine-5'-monophosphate disodium salt, 99+%
CAS: 6757-06-8 | C9H12N3Na2O8P | 367.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
Catalog number ALFJ63376.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCytidine-5'-monophosphate disodium salt
CAS6757-06-8
ColorWhite
Recommended StorageKeep cold
TSCANo