Spermine tetrahydrochloride, 99%, Thermo Scientific Chemicals
Spermine tetrahydrochloride, 99%, Thermo Scientific Chemicals
Spermine tetrahydrochloride, 99%, Thermo Scientific Chemicals
Spermine tetrahydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Spermine tetrahydrochloride, 99%, Thermo Scientific Chemicals
Binds to the polyamine modulatory site of NMDA
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number ALFJ63060.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSpermine tetrahydrochloride
CAS306-67-2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Spermine tetrahydrochloride, is used to precipitate DNA from low salt aqueous buffers. Spermine tetrahydrochloride was used to precipitate DNA for flow cytometric analysis from fresh frozen breast cancer tissue, archival tissue of salivary gland, renal and thyroid tumors2 and red blood cells of chicken.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Spermine tetrahydrochloride, is used to precipitate DNA from low salt aqueous buffers. Spermine tetrahydrochloride was used to precipitate DNA for flow cytometric analysis from fresh frozen breast cancer tissue, archival tissue of salivary gland, renal and thyroid tumors2 and red blood cells of chicken.
Solubility
Soluble in water (348.2 g/L at 20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic. Store under inert gas. Keep away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Benveniste and Mayer. Multiple effects of spermine on N-methyl-D-aspartic acid receptor responses of rat cultured hippocampal neurones. J.Physiol. 1993, 464, (2), 131-63.
  2. Munir et al. Polyamines modulate the neurotoxic effects of NMDA in vivo. Brain Res. 1993, 616, (1-2), 163-70.