Flunarizine dihydrochloride, 99%
Flunarizine dihydrochloride, 99%
Flunarizine dihydrochloride, 99%
Flunarizine dihydrochloride, 99%
Thermo Scientific Chemicals
Flunarizine dihydrochloride, 99%
CAS: 30484-77-6 | C26H28Cl2F2N2 | 477.42 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ62969.031 g
Catalog number ALFJ62969.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS698999-85-8
IUPAC Namesodium 1-[(2R,3R,4S,5R)-5-({[({[(2R,3R,4R,5R)-5-(6-amino-9H-purin-9-yl)-3-hydroxy-4-(phosphonooxy)oxolan-2-yl]methyl phosphonato}oxy)(oxido)phosphoryl]oxy}methyl)-3,4-dihydroxyoxolan-2-yl]-3-carbamoyl-1λ⁵-pyridin-1-ylium
Molecular FormulaC21H27N7NaO17P3
InChI KeyJNUMDLCHLVUHFS-QBMPJTFONA-M
SMILES[Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)(O)=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Assay>=97.0 % (NADP, mono-Na) (calc. from enzymatic
Water=<6.0 % (K.F.)
Ratio of Absorbances0.18 to 0.24 (A280/A260, pH 7.5)
Appearance (Color)White to light yellow
View more
A dual sodium/calcium channel blocker; a cerebral and peripheral vasodilator