Phosphate, 0.2M buffer soln., pH 7.2
Phosphate, 0.2M buffer soln., pH 7.2
Phosphate, 0.2M buffer soln., pH 7.2
Phosphate, 0.2M buffer soln., pH 7.2
Thermo Scientific Chemicals
Phosphate, 0.2M buffer soln., pH 7.2
CAS: 7558-80-7119.976 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Catalog number ALFJ62899.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Concentration0.2M buffer soln.
Molecular Weight (g/mol)119.976
Quantity1 L
PackagingPlastic Bottle
CAS7558-80-7
View more
Sodium phosphate monobasic is a reagent with very high buffering capacity widely used in molecular biology, biochemistry and chromatography. Useful in conjunction with Sodium Phosphate, Dibasic in the preparation of biological buffers. Used in many applications including the purification of antibodies. It is also used in HPLC. Phosphates are often used in foods and in water treatment. It is added in animal feed, toothpaste, and evaporated milk. It is used as a thickening agent and emulsifier.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
Ambient Temperatures