Ketoprofen, Thermo Scientific Chemicals
Ketoprofen, Thermo Scientific Chemicals
Ketoprofen, Thermo Scientific Chemicals
Ketoprofen, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ketoprofen, Thermo Scientific Chemicals
A non-steroidal anti-inflammatory drug (NSAID); also inhibits cyclooxygenases-1 and -2
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFJ62702.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialKetoprofen
CAS22071-15-4
Health Hazard 1H301
Health Hazard 2GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P264b-P270-P301+P310-P330-P501c
View more
It is used as a anti-inflammatory; analgesic and an antipyretic drug cause the hypothalamus to override an interleukin-induced increase in temperature. They act by blocking the synthesis of prostaglandins by inhibiting cyclooxygenase, which converts arachidonic acid to cyclic endoperoxides, precursors of prostaglandins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a anti-inflammatory; analgesic and an antipyretic drug cause the hypothalamus to override an interleukin-induced increase in temperature. They act by blocking the synthesis of prostaglandins by inhibiting cyclooxygenase, which converts arachidonic acid to cyclic endoperoxides, precursors of prostaglandins.
Solubility
Slightly soluble in chloroform and methanol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Osipova NA, T.D, et al. Ketoprofen (ketonal): a drug for preventing and treating postoperative pain.Anesteziologiia i reanimatologiia.,1999,671-74.
  2. Ting ST, et al. Effect of ketoprofen, lidocaine local anesthesia, and combined xylazine and lidocaine caudal epidural anesthesia during castration of beef cattle on stress responses, immunity, growth, and behavior.Journal of animal science.,2003,81(5), 1281-1293.