Xylazine hydrochloride, Thermo Scientific Chemicals
Xylazine hydrochloride, Thermo Scientific Chemicals
Xylazine hydrochloride, Thermo Scientific Chemicals
Xylazine hydrochloride, Thermo Scientific Chemicals
Xylazine hydrochloride, Thermo Scientific Chemicals
Agonist at the alpha-2 adrenergic receptor
Have Questions?
Catalog NumberQuantity
ALFJ62394.065 g
Catalog number ALFJ62394.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS10102-25-7
IUPAC Namedilithium(1+) hydrate sulfate
Molecular FormulaH2Li2O5S
InChI KeyRKGLUDFWIKNKMX-UHFFFAOYSA-L
SMILES[Li+].[Li+].O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Water Content (Karl Fischer Titration)12.9-16.4% (0.9-1.2 waters of hydration)
Assay (unspecified)≥98.5 to ≤101.5% (Dry wt. basis)
FormCrystalline powder
Agonist at the alpha-2 adrenergic receptor