Diosmin, Thermo Scientific Chemicals
Diosmin, Thermo Scientific Chemicals
Diosmin, Thermo Scientific Chemicals
Diosmin, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diosmin, Thermo Scientific Chemicals
CAS: 520-27-4 | C28H32O15 | 608.549 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFJ62073.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1124-33-0
IUPAC Name4-nitropyridin-1-ium-1-olate
Molecular FormulaC5H4N2O3
InChI KeyRXKNNAKAVAHBNK-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=[N+]([O-])C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)156.0-166.0?C
Appearance (Color)Pale yellow to yellow
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥96.0%
Diosmin exhibits anti-proliferative and anti-inflammatory. It is used in the treatment of venous disease, i.e., chronic venous insufficiency (CVI) and hemorrhoidal disease (HD).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in DMSO (50 mg/ml), water (122 mg/ml at 25°C), and ethanol (<1mg/mL at 25°C).
Notes
Protect from heat. Incompatible with heat and oxidizing agents. Store at 4°C.
RUO – Research Use Only
General References:
  1. Chung-Hsi Chou; Cheng-Wei Liu; Deng-Jye Yang; Yi-Hsieng Samuel Wu; Yi-Chen Chen. Amino acid, mineral, and polyphenolic profiles of black vinegar, and its lipid lowering and antioxidant effects in vivo. Food Chemistry.2003, 168,
  2. Oomaidurai Senthamizhselvan; Jeganathan Manivannan; Thangarasu Silambarasan; Boobalan Raja. Diosmin pretreatment improves cardiac function and suppresses oxidative stress in rat heart after ischemia/reperfusion. European Journal of Pharmacology.2009, 736,