trans-Zeatin, 97%, Thermo Scientific Chemicals
trans-Zeatin, 97%, Thermo Scientific Chemicals
trans-Zeatin, 97%, Thermo Scientific Chemicals
trans-Zeatin, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
trans-Zeatin, 97%, Thermo Scientific Chemicals
Plant growth hormone that stimulates cell division
Have Questions?
Catalog NumberQuantity
ALFJ61972.MF50 mg
Catalog number ALFJ61972.MF
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mg
Request bulk or custom format
Chemical Identifiers
CAS304680-35-1
IUPAC Name1-hexyl-3-methyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
Molecular FormulaC10H19F6N2P
InChI KeyYPWSSSRXUOQNMQ-UHFFFAOYSA-N
SMILESF[P-](F)(F)(F)(F)F.CCCCCCN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
NMR>=98.5 %
Infrared spectrumConforms
Refractive index1.4170 to 1.4200 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow to orange
Appearance (Form)Liquid
trans-Zeatin regulates cell division, development, and nutrient processing. It acts as a high-affinity ligand for a number of receptors, including arabidopsis histidine kinases (AHKs), specifically AHK3, which is important in mediating plant development.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
trans-Zeatin regulates cell division, development, and nutrient processing. It acts as a high-affinity ligand for a number of receptors, including arabidopsis histidine kinases (AHKs), specifically AHK3, which is important in mediating plant development.
Solubility
Soluble in DMSO and DMF
Notes
Store at -20°C. Incompatible with heat and oxidizing agents.
RUO – Research Use Only
General References:
  1. Iwona Żur; Ewa Dubas; Monika Krzewska; Piotr Waligórski; Michał Dziurka; Franciszek Janowiak. Hormonal requirements for effective induction of microspore embryogenesis in triticale (x Triticosecale Wittm.) anther cultures. Plant Cell Reports. 2015, 34, (1), 47-62.
  2. Robbin L Koenig; Roy O Morris; Joe C Polacco. tRNA is the source of low-level trans-zeatin production in Methylobacterium spp. Journal of Bacteriology. 2002, 184, (7), 1832-1842.