Karaya Gum
Thermo Scientific Chemicals
Karaya Gum
CAS: 9000-36-6
Have Questions?
Catalog NumberQuantity
ALFJ61844.36500 g
Catalog number ALFJ61844.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialKaraya Gum
CAS9000-36-6
ColorBrown to Yellow
Recommended StorageAmbient temperatures
EINECS Number232-539-4
View more
Used as a thickener and emulsifier