Sodium citrate, 0.5M buffer soln., pH 6.0
Sodium citrate, 0.5M buffer soln., pH 6.0
Thermo Scientific Chemicals
Sodium citrate, 0.5M buffer soln., pH 6.0
Have Questions?
Catalog NumberQuantity
ALFJ61815.AP500 mL
Catalog number ALFJ61815.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS998-40-3
IUPAC Nametributylphosphane
Molecular FormulaC12H27P
InChI KeyTUQOTMZNTHZOKS-UHFFFAOYSA-N
SMILESCCCCP(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥92.5%
FormLiquid
Sodium citrate, 0.5M buffer soln., pH 6.0 is used for RNA isolation. It aids in the detection of antigens in fixed tissue preparations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium citrate, 0.5M buffer soln., pH 6.0 is used for RNA isolation. It aids in the detection of antigens in fixed tissue preparations.
Solubility
Fully miscible in water.
Notes
Incompatible with heat and oxidizing agents.
RUO – Research Use Only
General References:
  1. Mostafa Mesbah; Usha Premachandran; William B. Whitman. Precise Measurement of the G+C Content of Deoxyribonucleic Acid by High-Performance Liquid Chromatography. Int J Syst Evol Microbiol.1989, 39, 159-167.
  2. Tomas Lindahl; Annika Andersson. Rate of chain breakage at apurinic sites in double-stranded deoxyribonucleic acid. Biochemistry.1972, 11 , (19), 3618-3623.