Sulindac
Sulindac
Sulindac
Thermo Scientific Chemicals
Sulindac
Have Questions?
Catalog NumberQuantity
ALFJ61772.065 g
Catalog number ALFJ61772.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS3520-42-1
IUPAC Namesodium 3,6-bis(diethylamino)-9-(2,4-disulfonatophenyl)-2,7-dimethyl-10λ⁴-xanthen-10-ylium
Molecular FormulaC29H33N2NaO7S2
InChI KeyXJENLUNLXRJLEZ-UHFFFAOYSA-M
SMILES[Na+].CCN(CC)C1=CC2=[O+]C3=CC(N(CC)CC)=C(C)C=C3C(C3=CC=C(C=C3S([O-])(=O)=O)S([O-])(=O)=O)=C2C=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown to dark brown
FormPowder
CommentSpecific absorbance @ Lambda max >1000
Loss on Drying≤15% (@ 105?C)
lambda-max (UV-Vis)561 - 566 nm in water
Sulindac inhibits the NF-κB pathway. Sulindac is a non-steroidal anti-inflammatory drug. Sulindac reduces the level and activity of several cyclin-dependent kinases and induces apoptosis. It is an antitumor agent. Sulindac is an inhibitor of Cox-1 and Cox-2.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase-1 and induces apoptosis in HT-29 cells.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C