XTT sodium salt
XTT sodium salt
XTT sodium salt
Thermo Scientific Chemicals
XTT sodium salt
CAS: 111072-31-2 | C22H16N7NaO13S2 | 673.52 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ61726.MD250 mg
Catalog number ALFJ61726.MD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS1013-88-3
IUPAC Namediphenylmethanimine
Molecular FormulaC13H11N
InChI KeySXZIXHOMFPUIRK-UHFFFAOYSA-N
SMILESN=C(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to yellow-brown
Refractive Index1.6150-1.6210 @ 20°C
Assay (GC)≥94.0%
StabilizerApprox. 0.02% w/w Hydroquinone
FormLiquid
Tetrazolium derivative useful in cancer research.XTT sodium salt is used in conjunction with phenazine methosulfate to screen human cancer cell lines. It is also used to study fungal cell damage. It is involved in testing antimicrobial susceptibility of staphylococci and in candida biofilm research. Further, it is used in an assay for cell viability and cell proliferation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetrazolium derivative useful in cancer research.XTT sodium salt is used in conjunction with phenazine methosulfate to screen human cancer cell lines. It is also used to study fungal cell damage. It is involved in testing antimicrob
RUO – Research Use Only
General References:
  1. Chew, S. Y.; Cheah, Y. K.; Seow, H. F.; Sandai, D.; Than, L. T. L. In vitro Modulation of Probiotic Bacteria on the Biofilm of Candida glabrata. Anaerobe 2015, 34, 132-138.
  2. Khurram, M.; Lawton, L. A.; Edwards, C.; Iriti, M.; Hameed, A.; Khan, M. A.; Khan, F. A.; Rahman, S. U. Rapid Bioassay-Guided Isolation of Antibacterial Clerodane Type Diterpenoid from Dodonaea viscosa (L.) Jaeq. Int. J. Mol. Sci. 2015, 16 (9), 20290-20307.