Alendronate sodium trihydrate, 97%, Thermo Scientific Chemicals
Alendronate sodium trihydrate, 97%, Thermo Scientific Chemicals
Alendronate sodium trihydrate, 97%, Thermo Scientific Chemicals
Alendronate sodium trihydrate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Alendronate sodium trihydrate, 97%, Thermo Scientific Chemicals
CAS: 121268-17-5 | C4H18NNaO10P2 | 325.122 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ61397.031 g
Catalog number ALFJ61397.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS16561-29-8
IUPAC Name(1S,2S,6R,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate
Molecular FormulaC36H56O8
InChI KeyPHEDXBVPIONUQT-RGYGYFBISA-N
SMILESCCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Appearance (Color)Colorless to white to light yellow
Appearance (Form)Film or solid
Alendronate sodium trihydrate is used as a bone resorption inhibitor, farnesyl diphosphate synthase inhibitor and CD45 protein tyrosine phosphatase inhibitor. A robust inhibitor of bone resorption that induces apoptosis. Alendronate inhibits the migration and invasion of PC-3 cells (sc-2220) by interfering with the mevalonate pathway. Additionally, Alendronate, Sodium Salt is an inhibitor of CD45RC and MMP.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Alendronate sodium trihydrate is used as a bone resorption inhibitor, farnesyl diphosphate synthase inhibitor and CD45 protein tyrosine phosphatase inhibitor. A robust inhibitor of bone resorption that induces apoptosis. Alendronate inhibits the migration and invasion of PC-3 cells (sc-2220) by interfering with the mevalonate pathway. Additionally, Alendronate, Sodium Salt is an inhibitor of CD45RC and MMP.
Solubility
Soluble in water at 10mg/ml
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Osama Hussein et. al. Breast cancer cells inhibit spontaneous and bisphosphonate-induced osteoclast apoptosis. Bone . 2011, 48, (2), 202-211 .
  2. Minakshi Asnani et. al. Ab initio structure determination of anhydrous sodium alendronate from laboratory powder X-ray diffraction data. Journal of Pharmaceutical Sciences. 2009, 98, (6), 2113-2121.