Acetate Buffer (1 M, pH 5.0)
Acetate Buffer (1 M, pH 5.0)
Acetate Buffer (1 M, pH 5.0)
Acetate Buffer (1 M, pH 5.0)
Thermo Scientific Chemicals
Acetate Buffer (1 M, pH 5.0)
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500mL
Catalog number ALFJ60964.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500mL
Request bulk or custom format
Specifications
CAS127-09-3
Concentration1M buffer soln.
Molecular FormulaC2H3NaO2
MDL NumberMFCD00012459
InChI KeyVMHLLURERBWHNL-UHFFFAOYSA-M
View more
For precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Acetate buffers for amine coupling of proteins to Biacore sensor chip surfaces. Studies on microbial pili suggest that sodium acetate suppressed K99 production in E. coli strains cultured on a minimal medium. Experiments have shown that when added to food waste composting systems, NaOAc may be effective at counteracting the adverse effects of organic acids produced in the composting process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
For precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Acetate buffers for amine coupling of proteins to Biacore sensor chip surfaces. Studies on microbial pili suggest that sodium acetate suppressed K99 production in E. coli strains cultured on a minimal medium. Experiments have shown that when added to food waste composting systems, NaOAc may be effective at counteracting the adverse effects of organic acids produced in the composting process.
Solubility
Soluble in water (246 g/L) at 20°C, and ethanol.
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from water, moisture.
RUO – Research Use Only
General References:
  1. Francis DH,; Ryan CJ,; Fritzemeier JD.Effect of sodium acetate on expression of K99 pili by Escherichia coli. Infect Immun. 1983, 41(3), 1368-9..
  2. Bingel M,; Lonnemann G,; Koch KM,; Dinarello CA,; Shaldon S. Enhancement of in-vitro human interleukin-1 production by sodium acetate.. Lancet. 1987, 1 (8523), 14-6.