(-)-Gallocatechin
(-)-Gallocatechin
(-)-Gallocatechin
Thermo Scientific Chemicals
(-)-Gallocatechin
CAS: 3371-27-5 | C15H14O7 | 306.27 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ60731.MA10 mg
Catalog number ALFJ60731.MA
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 mg
Request bulk or custom format
Chemical Identifiers
CAS41513-04-6
IUPAC Name1-bromo-4-chloro-2-nitrobenzene
Molecular FormulaC6H3BrClNO2
InChI KeyUKTIMFAJRPSNGR-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC(Cl)=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)66.0-72.0?C
Appearance (Color)Pale cream to cream to yellow to brown
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
Polyphenolic compound from green tea. Possesses free-radical scavenging ability
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Polyphenolic compound from green tea
  • Possesses free-radical scavenging ability
RUO – Research Use Only