(+)-Tubocurarine chloride pentahydrate, 98+%
(+)-Tubocurarine chloride pentahydrate, 98+%
(+)-Tubocurarine chloride pentahydrate, 98+%
(+)-Tubocurarine chloride pentahydrate, 98+%
Thermo Scientific Chemicals
(+)-Tubocurarine chloride pentahydrate, 98+%
Competitive, non-selective nicotinic acetylcholine receptor antagonist | CAS: 57-94-3 | C37H43Cl2N2O6+ | 682.659 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ60222.MC100 mg
Catalog number ALFJ60222.MC
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Request bulk or custom format
Chemical Identifiers
CAS1215564-15-0
IUPAC Name1-[4-(trifluoromethyl)pyrimidin-2-yl]-1H-pyrazole-4-sulfonyl chloride
Molecular FormulaC8H4ClF3N4O2S
InChI KeySADUDBJMXGGXDE-UHFFFAOYSA-N
SMILESFC(F)(F)C1=NC(=NC=C1)N1C=C(C=N1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream
Assay (Titration ex Chloride)≥94.0 to ≤106.0%
Assay (GC)≥94.0%
Melting Point (clear melt)92.0-105.0?C
FormPowder
Competitive, non-selective nicotinic acetylcholine receptor antagonist