Peroxidase, horseradish
Peroxidase, horseradish
Thermo Scientific Chemicals
Peroxidase, horseradish
Used in the oxidation of luminol to 3-aminophthalate. It is often used in conjugates to determine the presence of a molecular target, especially in the detection of specific protein in a western blot after conjugation with antibody.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
100 mg
Catalog number ALFJ60026.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS3147-14-6
IUPAC Name4-[2-(2-hydroxy-5-methylphenyl)hydrazin-1-ylidene]-3-oxo-3,4-dihydronaphthalene-1-sulfonic acid
Molecular FormulaC17H14N2O5S
InChI KeyASFVMSDYPYMUFL-UHFFFAOYSA-N
SMILESCC1=CC(NN=C2C(=O)C=C(C3=CC=CC=C23)S(O)(=O)=O)=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Black
Specific extinction E (1%/1cm)>=300 (610 nm, in buffer)
Appearance (Form)Crystalline powder
Suitability(for Ca + Mg determination) Passes test
Peroxidase, horseradish is used in the oxidation of luminol to 3-aminophthalate. It is often used in conjugates to determine the presence of a molecular target, especially in the detection of specific protein in a western blot after conjugation with antibody. Further, it is highly reactive toward human tumor cells.