Check Order Status
Bis-Tris, ≥98% (Dry Basis), Ultrapure, Thermo Scientific Chemicals
Element Search
Structure Search
Thermo Scientific Chemicals
Bis-Tris, ≥98% (Dry Basis), Ultrapure, Thermo Scientific Chemicals
CAS: 6976-37-0 | C8H19NO5 | 209.242 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
ALFJ12112.22
100 g
Plastic bottle
Catalog number
ALFJ12112.22
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS
6272-38-4
IUPAC Name
2-(benzyloxy)phenol
Molecular Formula
C13H12O2
InChI Key
CCZCXFHJMKINPE-UHFFFAOYSA-N
SMILES
OC1=CC=CC=C1OCC1=CC=CC=C1
View more
Specifications
Specification Sheet
Form
Liquid or viscous liquid
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless to pale brown or pale pink to red
Refractive Index
1.5885-1.5925 @ 20?C
RUO – Research Use Only