Ammonium Sulfate, ACS, ≥99%, Ultrapure, Thermo Scientific Chemicals
Ammonium Sulfate, ACS, ≥99%, Ultrapure, Thermo Scientific Chemicals
Ammonium Sulfate, ACS, ≥99%, Ultrapure, Thermo Scientific Chemicals
Ammonium Sulfate, ACS, ≥99%, Ultrapure, Thermo Scientific Chemicals
Ammonium Sulfate, ACS, ≥99%, Ultrapure, Thermo Scientific Chemicals
Used in purification of enzymes and other proteins by precipitation
Have Questions?
Catalog NumberQuantityPackaging
ALFJ11254.A11 kgPlastic bottle
Catalog number ALFJ11254.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS634-47-9
IUPAC Name2-chloro-4-methylquinoline
Molecular FormulaC10H8ClN
InChI KeyPFEIMKNQOIFKSW-UHFFFAOYSA-N
SMILESCC1=C2C=CC=CC2=NC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)53.0-59.0?C
Appearance (Color)White
Assay (GC)≥98.5%
Used in the purification of enzymes and other proteins by precipitation.