L-Dihydroorotic acid, 98%, Thermo Scientific Chemicals
L-Dihydroorotic acid, 98%, Thermo Scientific Chemicals
L-Dihydroorotic acid, 98%, Thermo Scientific Chemicals
L-Dihydroorotic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L-Dihydroorotic acid, 98%, Thermo Scientific Chemicals
CAS: 5988-19-2 | C5H6N2O4 | 158.113 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALFH60429.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS59-14-3
IUPAC Name5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC9H11BrN2O5
InChI KeyWOVKYSAHUYNSMH-RRKCRQDMSA-N
SMILESOC[C@H]1O[C@H](C[C@@H]1O)N1C=C(Br)C(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Related substances=<0.5 % (HPLC) 5-Bromouracil
Related substances=<0.2 % (HPLC) Each unknown impurity
Related substances=<1.0 % (HPLC) Total impurities
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder or crystals
View more
L-Dihydroorotic acid a metabolite.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Dihydroorotic acid a metabolite.
Solubility
Soluble in water (partly), and dimethyl formamide.
Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only
Charles S. Miller.; Janice T. Gordon.; Edward L. Engelhardt. Dihydroörotic Acid.J. Am. Chem. Soc.. 1953, 75 (23), 6086-6087.