Nalpha-Fmoc-O-tert-butyl-L-threonine, 98%, Thermo Scientific Chemicals
Nalpha-Fmoc-O-tert-butyl-L-threonine, 98%, Thermo Scientific Chemicals
Nalpha-Fmoc-O-tert-butyl-L-threonine, 98%, Thermo Scientific Chemicals
Nalpha-Fmoc-O-tert-butyl-L-threonine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nalpha-Fmoc-O-tert-butyl-L-threonine, 98%, Thermo Scientific Chemicals
CAS: 71989-35-0 | C23H27NO5 | 397.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number ALFH59808.04
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 g
Request bulk or custom format
Chemical Identifiers
CAS7783-90-6
IUPAC Namesilver(1+) chloride
Molecular FormulaAgCl
InChI KeyHKZLPVFGJNLROG-UHFFFAOYSA-M
SMILES[Cl-].[Ag+]
View more
SpecificationsSpecification SheetSpecification Sheet
Lead (Pb)=<10 ppm
Appearance (Color)White to pink-beige to grey
Iron (Fe)=<10 ppm
Appearance (Form)Powder
Copper (Cu)=<4 ppm