Diphenyl-2-pyridylphosphine, 98%, Thermo Scientific Chemicals
Diphenyl-2-pyridylphosphine, 98%, Thermo Scientific Chemicals
Diphenyl-2-pyridylphosphine, 98%, Thermo Scientific Chemicals
Diphenyl-2-pyridylphosphine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diphenyl-2-pyridylphosphine, 98%, Thermo Scientific Chemicals
CAS: 37943-90-1 | C17H14NP | 263.28 g/mol
Have Questions?
Catalog NumberQuantity
ALFH56029.065 g
Catalog number ALFH56029.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS25990-10-7
Molecular Formula(C6H8O6)n
SMILESO[C@@H]1[C@@H](O)[C@@H](O-*)[C@H](OC1-*)C(O)=O
MDL NumberMFCD00131972
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Identification (FTIR)Conforms
pH2.5-3.5 (1% suspension)
Appearance (Color)White to cream
Assay (unspecified)≥86.0% (dry wt. basis)
View more
It is a ligand for metal-catalyzed carbonylations, hydration, dehydrogenative coupling, carbostannylation, distannylation, and silylation. It is also employed as a reagent for Mitsunobu reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a ligand for metal-catalyzed carbonylations, hydration, dehydrogenative coupling, carbostannylation, distannylation, and silylation. It is also employed as a reagent for Mitsunobu reaction.
Solubility
Soluble in toluene. (almost transparency)
Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
RUO – Research Use Only
General References:
  1. Toshiyuki Oshiki.; Hitoshi Yamashita.; Kensuke Sawada.; Masaru Utsunomiya.; Kazunari Takahashi.; Kazuhiko Takai. Dramatic Rate Acceleration by a Diphenyl-2-pyridylphosphine Ligand in the Hydration of Nitriles Catalyzed by Ru(acac)2 Complexes. Organometallics. 2005, 24 (26),6287-6290 .
  2. Mehrak Kiankarimi.; Richard Lowe.; James R. McCarthy.; Jeffrey P. Whitten. Diphenyl 2-pyridylphosphine and di-tert-butylazodicarboxylate:Convenient reagents for the Mitsunobu reaction. Tetrahedron Lett. 1999, 40 (24),4497-4500 .