2-Nitrofluorene, 98%, Thermo Scientific Chemicals
2-Nitrofluorene, 98%, Thermo Scientific Chemicals
2-Nitrofluorene, 98%, Thermo Scientific Chemicals
2-Nitrofluorene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Nitrofluorene, 98%, Thermo Scientific Chemicals
CAS: 607-57-8 | C13H9NO2 | 211.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFH55744.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1189-71-5
IUPAC Name[(chlorosulfonyl)imino]methanone
Molecular FormulaCClNO3S
InChI KeyWRJWRGBVPUUDLA-UHFFFAOYSA-N
SMILESClS(=O)(=O)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with NaOH>=98.0 %
Appearance (Form)Liquid
Refractive index1.4450 to 1.4490 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow