Tri-n-butylphenyltin, 97%, Thermo Scientific Chemicals
Tri-n-butylphenyltin, 97%, Thermo Scientific Chemicals
Tri-n-butylphenyltin, 97%, Thermo Scientific Chemicals
Tri-n-butylphenyltin, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tri-n-butylphenyltin, 97%, Thermo Scientific Chemicals
CAS: 960-16-7 | C18H32Sn | 367.164 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number ALFH55596.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS34725-61-6
IUPAC Namesodium 3,5-dibromo-4-[3-(2,6-dibromo-4-hydroxyphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]benzen-1-olate
Molecular FormulaC19H9Br4NaO5S
InChI KeyKGANHHDZDYZJEM-UHFFFAOYNA-M
SMILES[Na+].OC1=CC(Br)=C(C(Br)=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=C(Br)C=C([O-])C=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Color ChangeYellow @ pH 3.0 to blue @ pH 4.6
Appearance (Color)Dark green to blue to brown to dark grey to black
Solution Test0.1% in water: Clear
It is applied in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is applied in chemical research.
Solubility
Sparingly Soluble (9.9E-5 g/L) (25°C)
Notes
Store at room temperature. Avoid strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Dietmar. Seyferth.; Michael A. Weiner. The Preparation of Organolithium Compounds by the Transmetalation Reaction. IV. Some Factors Affecting the Transmetalation Reaction. J. Am. Chem. Soc. 1962, 84 (3),361-364.
  2. Dietmar Seyferth.; Michael Weiner. Communications: Preparation and Some Reactions of Allyllithium. J. Org. Chem. 1959, 24 (9),1395-1396.