4-Fluoro-N-methylbenzylamine, 97%, Thermo Scientific Chemicals
4-Fluoro-N-methylbenzylamine, 97%, Thermo Scientific Chemicals
4-Fluoro-N-methylbenzylamine, 97%, Thermo Scientific Chemicals
4-Fluoro-N-methylbenzylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Fluoro-N-methylbenzylamine, 97%, Thermo Scientific Chemicals
CAS: 405-66-3 | C8H10FN | 139.173 g/mol
Have Questions?
Catalog NumberQuantity
ALFH54369.065 g
Catalog number ALFH54369.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS886-66-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Melting Point (clear melt)84.0-91.0°C