Tris(2-carboxyethyl)phosphine hydrochloride, 95%, 0.5M soln. in water, Thermo Scientific Chemicals
Tris(2-carboxyethyl)phosphine hydrochloride, 95%, 0.5M soln. in water, Thermo Scientific Chemicals
Tris(2-carboxyethyl)phosphine hydrochloride, 95%, 0.5M soln. in water, Thermo Scientific Chemicals
Tris(2-carboxyethyl)phosphine hydrochloride, 95%, 0.5M soln. in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tris(2-carboxyethyl)phosphine hydrochloride, 95%, 0.5M soln. in water, Thermo Scientific Chemicals
CAS: 51805-45-9 | C9H12O6P | 247.16 g/mol
Have Questions?
Catalog NumberQuantity
ALFH51864.AA5 mL
Catalog number ALFH51864.AA
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 mL
Request bulk or custom format
Chemical Identifiers
CAS139631-62-2
IUPAC Namecyclopropanesulfonyl chloride
Molecular FormulaC3H5ClO2S
InChI KeyPFWWSGFPICCWGU-UHFFFAOYSA-N
SMILESClS(=O)(=O)C1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
GC>=96.0 %
Tris(2-carboxyethyl)phosphine hydrochloride is widely used as a reducing agent to reduce disulfide bonds in proteins and trans-4,5-dihydroxy-1,2-dithiane. It is involved in the reduction of sulfoxides, sulfonyl chlorides, N-oxides and azides. It plays an important role in the tissue homogenization process for RNA isolation. It acts as a chelating agent and forms complexes with various heavy metal ions such as Zn(II), Cd(II), Pb(II), and Ni(II). Furthermore, it is useful in the labeling of cysteine residues with maleimides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tris(2-carboxyethyl)phosphine hydrochloride is widely used as a reducing agent to reduce disulfide bonds in proteins and trans-4,5-dihydroxy-1,2-dithiane. It is involved in the reduction of sulfoxides, sulfonyl chlorides, N-oxides and azides. It plays an important role in the tissue homogenization process for RNA isolation. It acts as a chelating agent and forms complexes with various heavy metal ions such as Zn(II), Cd(II), Pb(II), and Ni(II). Furthermore, it is useful in the labeling of cysteine residues with maleimides.
Solubility
Miscible with water and methanol.
Notes
Air sensitive. Store in cool place. Incompatible with strong oxidizing agents
RUO – Research Use Only
General References:
  1. La, M.; Feng, Y.; Yang, C.; Chen, C. Performances of p-Aminophenol Redox Cycling by Thiols and Tris (2-carboxyethyl) phosphine on Cysteamine-and 3-Mercaptopropionic acid-Covered Gold Electrodes. Int. J. Electrochem. Sci. 2014, 9, 6985-6992.
  2. Miralles, G.; Verdié, P.; Puget, K.; Maurras, A.; Martinez, J.; Subra, G. Microwave-mediated reduction of disulfide bridges with supported (Tris (2-carboxyethyl) phosphine) as resin-bound reducing agent. ACS Comb. Sci. 2013, 15 (4), 169-173.