Cyclohexylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals
Cyclohexylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals
Cyclohexylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals
Cyclohexylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals
CAS: 931-51-1 | C6H11ClMg | 142.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number ALFH51165.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS101-39-3
IUPAC Name(2Z)-2-methyl-3-phenylprop-2-enal
Molecular FormulaC10H10O
InChI KeyVLUMOWNVWOXZAU-CLFYSBASSA-N
SMILESCC(C=O)=CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear pale yellow to yellow
Free acid (titration)≤1.5%
Assay (GC)≥96.0%
Refractive Index1.6030-1.6080 @ 20?C
Cyclohexylmagnesium chloride is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF). It is used as a reagent for the introduction of the cyclohexyl group; metalation of CH-acidic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cyclohexylmagnesium chloride is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF). It is used as a reagent for the introduction of the cyclohexyl group; metalation of CH-acidic compounds.
Solubility
It reacts violently with water.
Notes
Air & moisture sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Yoshio Okamoto; Koji Ohta; Heimei Yuki. Asymmetric-selective polymerization of (RS)-α-methylbenzyl methacrylate with the cyclohexylmagnesium chloride-(---)- Sparteine system in toluene at -78°C. Macromolecules. 1978, 11, (4),724-727
  2. Jinhwa Lee; Heejin Kim; Jin Kun Cha. A new variant of the kulinkovich hydroxycyclopropanation. Reductive coupling of carboxylic esters with terminal olefins. J. Am. Chem. Soc. 1996, 118, (17),4198-4199