n-Butyllithium, 2.5M in hexane, packaged under Nitrogen in resealable AcroSeal™ bottles, Thermo Scientific Chemicals
n-Butyllithium, 2.5M in hexane, packaged under Nitrogen in resealable AcroSeal™ bottles, Thermo Scientific Chemicals
n-Butyllithium, 2.5M in hexane, packaged under Nitrogen in resealable AcroSeal™ bottles, Thermo Scientific Chemicals
n-Butyllithium, 2.5M in hexane, packaged under Nitrogen in resealable AcroSeal™ bottles, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Butyllithium, 2.5M in hexane, packaged under Nitrogen in resealable AcroSeal™ bottles, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
50 mL
Catalog number ALFH36949.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS55-21-0
IUPAC Namebenzamide
Molecular FormulaC7H7NO
InChI KeyKXDAEFPNCMNJSK-UHFFFAOYSA-N
SMILESNC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
HPLC>=98.5 %
Appearance (Form)Crystalline powder
Melting point125°C to 129°C