4,5-Dimethyl-o-phenylenediamine, 98%, many contain up to ca 15% water, Thermo Scientific Chemicals
4,5-Dimethyl-o-phenylenediamine, 98%, many contain up to    ca 15% water, Thermo Scientific Chemicals
4,5-Dimethyl-o-phenylenediamine, 98%, many contain up to    ca 15% water, Thermo Scientific Chemicals
4,5-Dimethyl-o-phenylenediamine, 98%, many contain up to    ca 15% water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4,5-Dimethyl-o-phenylenediamine, 98%, many contain up to ca 15% water, Thermo Scientific Chemicals
CAS: 3171-45-7 | C8H12N2 | 136.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFH32695.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material4,5-Dimethyl-o-phenylenediamine
Melting Point127°C to 129°C
CAS3171-45-7
Health Hazard 1H301-H312+H332-H315-H319
Health Hazard 2GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
4, 5-dimethyl-o-phenylenediamine used in the determination of aromatic aldehydes by high performance liquid chromatography after precolumn fluorescent derivatization. For the determination of vanillin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4, 5-dimethyl-o-phenylenediamine used in the determination of aromatic aldehydes by high performance liquid chromatography after precolumn fluorescent derivatization. For the determination of vanillin.
Solubility
Slightly soluble in water.
Notes
Air & Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, heat, light, acid chlorides, acid anhydrides.
RUO – Research Use Only
General References:
  1. Masatoki Katayama,; Yoshio Mukai,; Hirokazu Taniguchi. Fluorometric Determination of Vanillin with 4, 5-Dimethyl-o-phenylenediamine. Analytical Sciences. 1987, 3(4), 369-372.
  2. Glen R. Gale,; Loretta M. Atkins,; Ernest M. Walker Jr.,; Alayne B. Smith,; Sandra J. Meischen. Antitumor Action of Dichloro (4,5-dimethyl-o-phenylenediamine-N,N') platinum (II)1. Exp Biol Med (Maywood). 1973, 142(4), 1349-1354.