Furan-2,5-dicarboxylic acid, 98%, Thermo Scientific Chemicals
Furan-2,5-dicarboxylic acid, 98%, Thermo Scientific Chemicals
Furan-2,5-dicarboxylic acid, 98%, Thermo Scientific Chemicals
Furan-2,5-dicarboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Furan-2,5-dicarboxylic acid, 98%, Thermo Scientific Chemicals
CAS: 3238-40-2 | C6H4O5 | 156.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFH28718.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS107-13-1
IUPAC Nameprop-2-enenitrile
Molecular FormulaC3H3N
InChI KeyNLHHRLWOUZZQLW-UHFFFAOYSA-N
SMILESC=CC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥99.0%
CommentStabilizer: ca 40 ppm 4-methoxyphenol