Lithium bis(trifluoromethylsulfonyl)imide, 98+%, Thermo Scientific Chemicals
Lithium bis(trifluoromethylsulfonyl)imide, 98+%, Thermo Scientific Chemicals
Lithium bis(trifluoromethylsulfonyl)imide, 98+%, Thermo Scientific Chemicals
Lithium bis(trifluoromethylsulfonyl)imide, 98+%, Thermo Scientific Chemicals
Lithium bis(trifluoromethylsulfonyl)imide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Lithium bis(trifluoromethylsulfonyl)imide, 98+%, Thermo Scientific Chemicals
CAS: 90076-65-6 | C2F6LiNO4S2 | 287.075 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number ALFH27307.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS7440-47-3
IUPAC Namechromium
Molecular FormulaCr
InChI KeyVYZAMTAEIAYCRO-UHFFFAOYSA-N
SMILES[Cr]
View more
SpecificationsSpecification SheetSpecification Sheet
FormChunks/pieces, 2-15 mm (0.08-0.59 in.)
Total Metal Impurities≤0.004%
Lithium bis(trifluoromethylsulfonyl)imide is used in the preparation of chiral imidazolium salt through an anion metathesis of the corresponding triflate organic electrolyte-based lithium batteries. It finds application in the preparation of rare-earth Lewis acid catalysts. It is also useful in primary and secondary lithium cells using organic liquid electrolytes and polymer batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Lithium bis(trifluoromethylsulfonyl)imide is used in the preparation of chiral imidazolium salt through an anion metathesis of the corresponding triflate organic electrolyte-based lithium batteries. It finds application in the preparation of rare-earth Lewis acid catalysts. It is also useful in primary and secondary lithium cells using organic liquid electrolytes and polymer batteries.
Solubility
Soluble in water.
Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Motomatsu, J.; Kodama, H.; Furukawa, T.; Tominaga, Y. Dielectric Relaxation Behavior of a Poly (ethylene carbonate)-Lithium Bis-(trifluoromethanesulfonyl) Imide Electrolyte. Macromol. Chem. Phys. 2015, 216, (15), 1660-1665.
  2. Pehlivan, I. B.; Granqvist, C. G.; Niklasson, G. A. Ion conduction mechanism of nanocomposite polymer electrolytes comprised of polyethyleneimine-lithium bis (trifluoromethylsulfonyl) imide and silica. Electrochim. Acta 2014, 119, 164-168.